missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Adapter Fittings, Sanitary Clamp to Hose Barb, Straight

Maintain sanitary conditions while quickly connecting to tubing.

Brand:  Masterflex™ 31805-26

Code : A0

Additional Details : Weight : 0.00450kg

 View more versions of this product

Product Code. 11827533

  • £173.00 / Each
Estimated Shipment: 29-10-2024
to see stock.
Add to basket

Description

Description

  • Allowing for easy assembly and disassembly of sanitary set-ups, these adapters are suitable for use connecting a Tri-Clamp™ fitting to flexible tubing with the hose barb adapter
  • Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving
  • Clamps and gaskets required for a complete system are sold separately
Specifications

Specifications

Sanitary Barbed Adapter
Polypropylene
-9°C
82°C
Autoclavable
Fits 3/4 in. Clamp Size x 3/8 in. Tubing I.D.
1 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers