Learn More
Vitlab™ Polypropylene Scoop
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ Polypropylene Scoops are lighter than metal, don’t dent and feature good chemical resistance. Also suitable as weighing scoops.
Brand: Vitlab™ 39494
This item is not returnable.
View return policy
Description
- White polypropylene scoop
- With precision formed filling edge and comfortable, stable handle
- Easily readable volume quantities on the upper side of the handle
Specifications
Measuring Scoop | |
25 mL | |
For contact with food |
Polypropylene | |
White | |
135 mm |