Learn More
Macherey-Nagel™ Polypropylene Screw Caps with Hole
Polypropylene screw closures for sample storage with an analytically pure Silicone/PTFE septum that is firmly connected to the cap.
Brand:  Macherey-Nagel™ 702113
Product Code. 15914880
  • £21.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Closed top
  • White
  • Whie silicone/blue PTFE pre-fitted septa
Specifications
Specifications
Screw Caps with Silicone PTFE Septa
Polypropylene
Sample Storage
1.3 mm (Septa)
White
White Silicone/Blue PTFE
Temp. Resistance Septa -60°C up to 200°C - However, PP cap only up to 120°C/Analytical Purity - High
Screw Cap