Learn More
Macherey-Nagel™ Polypropylene Screw Caps with Hole
Polypropylene closures with red rubber/FEP septum for routine analysis and a fair price-performance-ratio. Easy and safe needle penetration. Compatible with Agilent, Knauer autosamplers and GC/HPLC applications.
Brand:  Macherey-Nagel™ 702082
Product Code. 12725232
  • £16.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Green color
  • Red rubber/colorless FEP septum
  • For GC and HPLC applications
  • Pre-assembled
Specifications
Specifications
Red Rubber/Colorless FEP
Green
Red Rubber/Colorless FEP
Temp. Resistance -40°C up to 110°C/Analytical Purity - Medium/Medium Hard to Penetrate/Medium Resealability
Agilent and Knauer Autosamplers
PP Screw Cap with Hole and Pre-Fitted Septa
Polypropylene
GC and HPLC
Screw Cap