Learn More
Fisherbrand™ Polypropylene Spheres for Bath
A useful alternative to a lid, these spheres minimize evaporation and heat loss while allowing easy access to vessels in the bath.
Brand: Fisherbrand™
Additional Details : Weight : 0.40000kg
Description
- Particularly useful for tall vessels
- Lid or layer of polypropylene spheres should be used between operating temperatures 60°C and 100°C, and below room temperature
Specifications
Cover | |
Polypropylene | |
300 Spheres | |
0.75 kg |