missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ Polypropylene Stackable Beaker
Use these stackable beakers for a variety of general lab applications. VITLAB™ Polypropylene Stackable Beakers feature a stable handle and easily readable, printed black scale on both sides. Volume is equally visible for left and right handers.
£107.00 - £145.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10261232
|
Vitlab™
483941 |
2000 mL |
£107.00
Pack of 12 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10397852
|
Vitlab™
484941 |
3000 mL |
£145.00
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
- Highly transparent
- To preserve markings, cleaning at no higher than 60°C is recommended
- Suitable for foodstuffs
Specifications
Polypropylene |