Learn More
Brand™ Polypropylene Stackable Tip Box
GREENER_CHOICE
Includes tip tray. BRAND™ Polypropylene Stackable Tip Box comes empty. Filter and pipet tips sold separately.
£6.10 - £8.10
Specifications
Product Type Empty Tip Box
Material Polypropylene
Stackable Yes
For Use With (Equipment) Pipet and Filter Tips
No. per Pack 1 Pack
View More Specs
Products 4
Product Code Brand Holds Color Price Quantity & Availability  
Product Code Brand Holds Color Price Quantity & Availability  
15134997
GREENER_CHOICE
View Documents Promotion Details
Brand™
732992
Pipette Tips (200 μL) Yellow
£7.35
Each
In Stock
to see stock.
Edge
Add to basket
15144997
GREENER_CHOICE
View Documents Promotion Details
Brand™
732996
Pipette Tips (1000 μL) Blue
£8.10
Each
In Stock
to see stock.
Edge
Add to basket
15124997
GREENER_CHOICE
View Documents Promotion Details
Brand™
732990
Pipette Tips (50 μL) Gray
£7.00
Each
In Stock
to see stock.
Add to basket
15154997
GREENER_CHOICE
View Documents Promotion Details
Brand™
732994
Pipette Tips (300 μL) Green
£6.10
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Manufactured from polypropylene
  • Includes tip tray
  • Empty, for filter and pipet tips
Specifications
Specifications
Empty Tip Box
Yes
1 Pack
Polypropylene
Pipet and Filter Tips
1 Pack
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Brand™ Polypropylene Stackable Tip Box