Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Variously sized and shaped tubing connectors. Ismatec™ Standard Tubing Connectors, Plastic are available in different plastic compositions.
Brand:  Ismatec™ ISM510
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10505373
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifications
Specifications
Standard Tubing Connector
For 3.0mm Tubing I.D.
3.0mm O.D.
Acetal Resin (POM, Polyoxymethylene)
10/Pk.
T-shape