Learn More
Fisherbrand™ Polypropylene Storage Boxes with lid, Autoclavable, for EPA-Vials
Temperature resistant from -80° up to +100°.
£41.55 - £42.65
Product Code | Brand | Dimensions (L x W x H) | For Use With (Equipment) | No. of Places | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | For Use With (Equipment) | No. of Places | Price | Quantity & Availability | |||
12652465
|
Fisherbrand™
28 21 2429 |
130 x 130 x 105 mm | For 30ml, 40ml EPA vials | 10 cavities |
£42.65
Each |
In Stock
Log in to see stock. |
|||
12652505
|
Fisherbrand™
28 21 2428 |
130 x 130 x 80 mm | For 20ml EPA vials | 16 cavities |
£41.55
Each |
In Stock
Log in to see stock. |
|||
Description
- Storage Box
- Includes cover
- Feature alphanumeric indexing for easier vial identification
- Stackable for efficient storage
Specifications
Violet |