Learn More
BRAND™ Polypropylene Thread Adapters
Shop All Brand Products
Click to view available options
For Use With (Equipment):
For bottle thread GL 22
For bottle thread GL 24-25
For bottle thread GL 30
For bottle thread S 40
For bottle thread S 54
For bottle thread S 60
For ground joint NS 24/29
Unit Size:
Each
Description
- Thread adapter, Polypropylene (PP), outer thread GL 45, for bottle thread S 40
- Thread Adapters for BRAND™ Bottle Top Dispensers, Polypropylene, 45/40 mm
- For use with Bottle Thread 40 S, Dispensette, Seripettor, Titrette and QuikSip Bottletop Diluters and Dispensers
- The Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use
- Ideal for use with Dispensetter bottletop dispensers
- These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND™ bottletop dispensers and burettes
- These thread adapters help to maximize the compatibility of bottletop dispensers with most reagent bottles
Specifications
Specifications
| Product Type | Thread Adapter |
| Material | Polypropylene |
| For Use With (Equipment) | For bottle thread GL 24-25 |
| Quantity | 1 Ea. |
| Thread Size | 32 GL |