Learn More
BRAND™ Polypropylene Thread Adapters
These Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use and are ideal for use with Dispensetter bottletop dispensers.
Brand:  BRAND™ 704430
 View more versions of this product
Product Code. 10592423
  • £32.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Thread adapter, Polypropylene (PP), outer thread GL 45, for bottle thread S 40
  • Thread Adapters for BRAND™ Bottle Top Dispensers, Polypropylene, 45/40 mm
  • For use with Bottle Thread 40 S, Dispensette, Seripettor, Titrette and QuikSip Bottletop Diluters and Dispensers
  • The Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use
  • Ideal for use with Dispensetter bottletop dispensers
  • These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND™ bottletop dispensers and burettes
  • These thread adapters help to maximize the compatibility of bottletop dispensers with most reagent bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thread Adapter
Polypropylene
For bottle thread S 54
Outer thread GL 45