Learn More
Ark Plas Products™ Polypropylene Threaded Male Luer
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Polypropylene Threaded Male Luers feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
Brand:  Ark Plas Products™ LGX86-PP0
Product Code. 11771976
  • £77.90 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Natural polypropylene luers
  • Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • Molded in virgin, lot-traceable material
  • Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Specifications
Autoclavable
-17°C
1/4-28 UNF
Natural
Flexible tubing or threaded ports
25/Pk.
1/4-28 UNF
135°C