Learn More
Fisherbrand™ Polypropylene Transparency Makro Pipet Tips
Use these pipet tips with Gilson™ Pipetman™/Neo Pipets. Fisherbrand™ Polypropylene Transparency Makro Pipet Tips feature uniform high quality and precision, a perfect fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.61200kg
Product Code. 11846943
  • £41.40 / Pack of 250
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
Designed for use when transferring large sample volume. Advanced moulding technology ensures tips are straight. Alternative option to serological pipettes. Non-sterile, certified RNase, DNase, DNA and pyrogen free.
Specifications
Specifications
1000 to 5000 μL
Natural
Makro
Non-sterile
For Use When Transferring Large Sample Volume
Polypropylene