missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polypropylene Utility Funnel

Designed for easy, economical funneling

Brand:  Cole-Parmer™ 06122-10

Product Code. 11775328

  • £37.50 / Pack of 12
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene utility funnel; 2 oz.
  • Ideal for general use or powders
  • Available in packs of 12
Specifications

Specifications

70mm
10mm
68 mm
2.75 in.
2 oz.
2.677 in.
70 mL
Polypropylene
White
2.677 in.
68mm
Utility Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers