missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Protein LoBind Microplate
For troublesome samples where protein concentrations are low, the Eppendorf LoBind plates ensure maximal sample recovery - protecting enzyme activity and facilitating downstream analysis.
£798.18 - £2300.00
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polypropylene |
No. of Wells | 384 |
Well Shape | Conical Bottom (V-well) |
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
11568653
|
Eppendorf™
0030628306 |
240 |
£2300.00
Pack of 240 |
In Stock
Log in to see stock. |
|||||
10702074
|
Eppendorf™
0030624300 |
80 |
£1025.00
Pack of 80 |
In Stock
Log in to see stock. |
|||||
10748483
|
Eppendorf™
0030524101 |
40 |
£798.18
Pack of 40 |
In Stock
Log in to see stock. |
|||||
10583984
|
Eppendorf™
0030528107 |
120 |
£2090.00
Pack of 120 |
In Stock
Log in to see stock. |
|||||
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- High-contrast Unique OptiTrack™ matrix: up to 30 % faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface ensure reliable sealing in plates
Specifications
Standard | |
Polypropylene | |
Conical Bottom (V-well) |
16 x 24 | |
384 |