missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Polypropylene Wadless Screw Caps
Click to view available options
Color:
Natural
White
Closure Type:
Wadless Screw Cap
Description:
Thread style: R3/20
Thread style: R3/22
Thread style: R3/24
Thread style: R3/28
Thread style: R3/33
Thread style: R3/38
Thread style: R3/48
Thread style: R3/51
Thread style: R3/58
Thread style: R3/63
Thread style: R3/83
Thread style: R6/31
Description
- For medical usages
- White polypropylene
- Softer texture due to white pigmentation
Specifications
Specifications
Color | White |
Description | Thread style: R3/38 |
Material | Polypropylene |
Quantity | 50/PK |
For Use With (Equipment) | For BTF-425, 430, 440, 600, 610, 625, 630 series bottles |
Closure Type | Wadless Screw Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'