Learn More
Fisherbrand™ Polypropylene Yellow Graduated Beveled Pipet Tip
Product Code. 11517452 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Pack of 1000
Product Code. 11517452
Only null left
Add to basket
Edge
Add to basket
Use these pipet tips with all commercially available pipets. Fisherbrand™ Polypropylene Yellow Graduated Beveled Pipet Tips feature uniform high quality and precision, a perfect fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
Specifications
Volume (Metric) 1 to 200 μL
Color Yellow
Autoclavable Autoclavable
Graduated Yes
Material Polypropylene