missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 40, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Polysorbate 40 has been used in a study to assess the determination of iodide in urine based on a chemiluminescence system. It has also been used in a study to investigate the delivery of astaxanthin and canthaxanthin to HepG2 cells. Also used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.09 |
Solubility Information | Fully miscible with water. Soluble in isopropyl alcohol (100mg/ml),and xylene. Insoluble in propylene glycol,and mineral oil. |
Quantity | 500 mL |
Refractive Index | 1.47 |
Chemical Name or Material | Polysorbate 40 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'