missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80

Product Code. p-2692440 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10389150

Brand: Thermo Scientific Acros 278632500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Synonym Polyoxyethylene(20)sorbitan monooleate

Specifications

Color Yellow to Amber
Content And Storage Indoors +20°C
Density 1.0800 g/mL
Quantity 250 mL
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL Number MFCD00082107
Packaging Type Glass bottle
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Molecular Weight (g/mol) 604.82
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil
Form Viscous Liquid
Chemical Name or Material Polysorbate 80
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.