Learn More
Poulten Graf™ Polystop Volac Stopper
Brand:  Poulten Graf™ L189/D
 View more versions of this product
Product Code. 12328259
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
For reagent bottles
16/16
Polystop Stopper
20/Pk.
Polypropylene