missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Polystyrene 1536-Well Microplates
Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
£1240.63 - £1943.19
Specifications
Format | Standard |
---|---|
Array | 32 x 48 |
Color | Black |
Material | Polystyrene |
No. of Wells | 1536 |
Product Code | Brand | Volume (Metric) Working | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Working | For Use With (Application) | Price | Quantity & Availability | ||||
11359514
|
Corning™
3838 |
10 μL | Fluorescent, luminescent assays |
£1943.19
Pack of 50 |
In Stock
Log in to see stock. |
||||
11373725
|
Corning™
3726BC |
Up to 8 μL | Supports expansion of undifferentiated stem cells |
£1240.63
Pack of 50 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
- Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
- Recommended working volume up to 8 μL
- Round well bottom for reduced air entrapment and improved CV values and Z factor
- Raised well bottom for higher sensitivity
- Flood reservoir on four sides to reduce instrument contamination
- Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Specifications
Standard | |
Black | |
1536 | |
12.8 μL | |
50 |
32 x 48 | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
50/Pk. |