Learn More
Corning™ Polystyrene Adapter Applicator
Applications include: PCR clean-up; dye-terminator clean-up and separation/filtration assays
Brand:  Corning™ 3507
Code : 66
Additional Details : Weight : 5.39774kg
Product Code. 10062521
  • £2695.57 / Each
Estimated Shipment: 25-11-2024
to see stock.
Add to basket
Description
Description
With a well volume of 360μL the plate body is made of white polystyrene with a K-Resin™ insert. The standard membrane consists of a low protein binding, hydrophilic 0.2μm PVDF (Polyvinylidene Fluoride). Another feature is the individual filter discs. These are encapsulated by a special over-moulding process that ensures the integrity of each well and eliminates liquid cross contamination, while the optimised nozzle design prevents cross-contamination of the filtrate. The plate has the standard industry (SBS) footprint, so is compatible with most automation, liquid handling and detection equipment. Main applications include: PCR* clean-up; dye-terminator clean-up and separation/filtration assays.
Specifications
Specifications
Hydrophilic 0.2μm Polyvinylidene Fluoride
White
Polystyrene
360 μL
Each
8 x 12
PCR clean-up; dye-terminator clean-up and separation/filtration assays
96
PCR clean-up, dye-terminator clean-up and separation/filtration assays