Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Made from anti-static, high impact polystyrene, these spill trays are ideal in most research, scientific, laboratory and other environments.
Brand:  Clark Scientific™ AT2-PK25
 View more versions of this product
Product Code. 12948837
  • £477.00 / Pack of 25
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from anti-static, high impact polystyrene
  • Sufficient depth to safely contain even the largest liquid spills
  • Wipe clean for repeated use
Specifications
Specifications
Polystyrene
25/Pk.
450 mm
Spill Tray
700 mm