Learn More
Sampling Systems™ SteriWare™ Disposable PharmaScoop™
Manufactured in a class 100,000 cleanroom to ensure cleanliness. Sampling Systems™ Polystyrene Disposable Pharma Scoops are individually wrapped and do not require expensive cleaning or cleaning validation.
£66.15 - £211.00
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
17458863
![]() |
Sampling Systems™
8045A-1000 |
1000 mL | Non-sterile |
£90.65
Pack of 25 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
17478863
![]() |
Sampling Systems™
8045A-2500 |
2.5 L | Non-sterile |
£66.15
Pack of 15 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
17498863
![]() |
Sampling Systems™
8045A-500 |
500 mL | Non-sterile |
£148.00
Pack of 50 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
17488863
![]() |
Sampling Systems™
8045A-2500S |
2.5 L | Sterile |
£112.00
Pack of 15 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
12534366
![]() |
Sampling Systems™
8045A-60S |
60 mL | Sterile |
£114.00
Pack of 100 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
17408873
![]() |
Sampling Systems™
8045A-500S |
500 mL | Sterile |
£176.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
17468863
![]() |
Sampling Systems™
8045A-1000S |
1000 mL | Sterile |
£132.00
Pack of 25 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
13225220
![]() |
Sampling Systems™
8045A-125S |
125 mL | Sterile |
£211.00
Pack of 100 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
16466840
![]() |
Sampling Systems™
8045A-250S |
250 mL | Sterile |
£211.00
Pack of 85 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
12732437
|
Sampling Systems™
8045A-250S |
2.5 L | Non-sterile | N/A | ||||||
13255220
![]() |
Sampling Systems™
8045A-30S |
30 mL | Sterile |
£112.00
Pack of 100 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
15809431
![]() |
Sampling Systems™
8045A-60 |
60 mL | Non-sterile |
£88.00
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
15879421
![]() |
Sampling Systems™
8045A-125 |
125 mL | Non-sterile |
£112.00
Pack of 100 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
15889421
|
Sampling Systems™
8045A-250 |
250 mL | Non-sterile | N/A | ||||||
16456840
![]() |
Sampling Systems™
8045A-250 |
250 mL | Non-sterile |
£133.00
Pack of 85 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
15899421
![]() |
Sampling Systems™
8045A-30 |
30 mL | Non-sterile |
£84.45
Pack of 100 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
Description
Key Features:
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Also available in HDPE, PP and other colors
Why use a SteriWare™ Disposable PharmaScoop™?
- GMP Correct - It is Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible. In accordance with this, we have positioned the ergonomic handle high up on the scoop.
- Cleanroom Manufacture - All SteriWare™ PharmaScoops™ are moulded, inspected and packed in a Class 100,000 Cleanroom.
- No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
- No Cleaning - Unlike a reusable scoop, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
- High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
- Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificate of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
- Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
- Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Polystyrene | |
Single-use | |
White |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable PharmaScoop™