missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polystyrene Disposable Test Tubes

Manufactured with polystyrene for superb clarity. Kartell™ Polystyrene Disposable Test Tubes are economical and durable.

Brand:  Kartell™ 8830800

 View more versions of this product

Product Code. 11352904

  • £63.45 / Pack of 1000
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured with polystyrene for superb clarity. Kartell™ Polystyrene Disposable Test Tubes are economical and durable.

  • Single use eliminates contamination from carry over or absorption
  • Excellent for sensitive batch work or low speed centrifugation
  • Color coded caps available separately
Specifications

Specifications

CE IVD
1300 x g
55 mm
Single-use
Non-sterile
Coagulometer Tube
Clear
3 mL
Polystyrene
1000 Pack
11.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers