missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polystyrene Drying Racks

Features 72 place support that can accomodate items with neck bore in excess of 15mm. Kartell™ Polystyrene Drying Racks deliver a sturdy, durable and adjustable solution for drying items in the laboratory.

Brand:  Kartell™ 0021300

171.52 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10182471

  • £204.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Features 72 place support that can accomodate items with neck bore in excess of 15mm. Kartell™ Polystyrene Drying Racks deliver a sturdy, durable and adjustable solution for drying items in the laboratory.

  • Molded high impact polystrene backplate provides strength and prevents rot, stain and rust
  • Close ended peg sockets eliminate leakage and potential consequent biohazards
  • Seamless drip channel collects waste; drains via supplied drain tube
  • 72 push in pegs supplied, easily removed for cleaning or to accomodate odd shaped items
  • Replacement pegs available
  • Racks can be joined with supplied plugs to create additional drying space
Specifications

Specifications

72 push in pegs, 11 small pegs, joining plugs, mounting kit, draining kit (connector and tubes), and fixing template
Polystyrene
Supplied discharging tube: I.D.: 11mm, O.D.: 13mm
1 Each
0.2 kg
Drainer
72 pegs accomodate items with neck bore in excess of neck bore in excess of 15mm, 6mm pegs (included) accomodate small diameter items, pegs can be removed to accomodate differently shaped objects
1 Each
450L x 110D x 630mmH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Polystyrene Drying Racks > Height: 630mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.