Learn More
Technical Service Consultants™ Polystyrene L-shaped Spreader
Click to view available options
Quantity:
1000/Pk.
500 Pk.
Unit Size:
Pack of 1000
Pack of 500
Description
- Polystyrene
- Three year shelf life
- Ideal for bacterial counts
- L shaped plastic (firm) peel pouch
- Hockey Stick Shape
Specifications
Specifications
| Quantity | 500 Pk. |
| Shape | L-Shape |
| Material | Polystyrene |
| Product Type | Spreader |
| Sterility | Sterile |