Learn More
Corning™ Polystyrene Microplates
GREENER_CHOICE
Designed for in vitro diagnostic assays
£740.00 - £1344.54
Specifications
Material Polystyrene
View More Specs
Products 3
Product Code Brand Total Volume (Metric) Well Surface Treatment Price Quantity & Availability  
Product Code Brand Total Volume (Metric) Well Surface Treatment Price Quantity & Availability  
10022551
View Documents Promotion Details
Corning™
2510
360 μL Sulfhydryl This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10424872
GREENER_CHOICE
View Documents Promotion Details
Corning™
2482
190 μL Non-treated
£768.31
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
15222319
View Documents Promotion Details
Corning™
3831BC
0.0128 mL Non-treated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.
  • Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
  • Recommended working volume of 50μL to 125μL
  • Designed for in vitro diagnostic assays
  • Untreated surface is hydrophobic in nature
  • Color coded strips
  • Bar coding available on request
Maleimide surface covalently couples to sulfhydryl groups (via SH moieties) and are ideal for assays requiring site directed orientation of a particular biomolecule, especially antibodies. Minimal surface preparation is required. Individually packed.
Specifications
Specifications
Polystyrene