missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millicell-24 Cell Culture Insert Plate
Millicell cell culture insert plates are a patented device designed to support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2.
Brand: Merck PSST010R5
Code : 71
Additional Details : Weight : 0.01000kg
Description
- Support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2
- Designed for maximum user convenience, easier pipetting and basolateral access
- Optimised to grow and sustain high integrity cell monolayers
- Plates have feet to elevate the plate above the work surface when disassembled from the feeder tray, reducing monolayer contamination
- Can be used manually or with automated cell seeding, feeding and washing systems
- Can be used with either a single-well or multi-well feeding tray
- Tear drop-shape receiver wells eliminate air bubbles as plates are assembled
Specifications
ISO 9001 | |
Not Autoclavable | |
0.7 cm2 (Filter) | |
Rectangular | |
Cell Culture Insert | |
Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport, Permeability | |
5 Pk. | |
5.0 μm | |
128 mm | |
26.2 mm |
Cell Culture Insert Plate, Isopore™ Media | |
Clear | |
Polycarbonate | |
Sterile | |
24 | |
24-well Filter Plate, 24-well Receiver Plate, Lid | |
Bulk Packed | |
50°C (max.) | |
84.8 mm | |
9.5 mm (Filter) |