missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polystyrene Petri Dishes

Product Code. p-8000232
Click to view available options
Dimensions (Dia. x H):
35 x 10 mm
60 x 15 mm
90 x 15 mm
94 x 15 mm
94 x 16 mm
100 x 20 mm
120 x 17 mm
145 x 20 mm
Sterility:
Non-sterile
Sterile
Vent:
No
Yes
Quantity:
120 Pack
240 Pk.
360 Pack
420 Pack
480 Pack
560 Pack
600 Pack
740 Pack
Unit Size:
Pack of 120
Pack of 240
Pack of 360
Pack of 420
Pack of 480
Pack of 560
Pack of 600
Pack of 740
This item is not returnable. View return policy

Product Code. 10470613

Brand: Greiner Bio-One 632180

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material Polystyrene
Shape Round
Sterility Non-sterile
Quantity 480 Pack
Vent No
Dimensions (Dia. x H) 94 x 16 mm
Height (Metric) 16 mm
Diameter (Metric) 94 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.