missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Polystyrene Sterile Single Wrapped Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
Brand: Heathrow Scientific™ 120750
146.11 GBP valid until 2024-12-31
Use promo code "21757" to get your promotional price.
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sterile, sealed in bags and electron beam sterilized
- DNase, RNase and Pyrogen free
- Sealed in easy-to-open bags
- Quality certificate available on request
- Disposable
Specifications
1.9 oz. | |
55 mL | |
Polystyrene | |
Reagent Reservoir | |
1/Bag, 100 Bags/Pk. | |
1 | |
Convoluted |
White | |
4 x 1.10 x 1.20 in. | |
Sterile | |
Single Wrapped | |
Not Autoclavable | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction