Learn More
Merck Millipore Millex™ LG Syringe Filter with Hydrophilic LCR PTFE Membrane
Designed for sample preparation and low protein binding fine particle removal from solvents, aqueous and organic solutions. Millex™ LG Syringe Filters with Hydrophilic LCR PTFE Membranes are for non-sterile filtration.
Description
Description
  • 0.2 μm pore size
  • Female Luer-Lok/Male slip
  • Applications include analytical sample prep