missing translation for 'onlineSavingsMsg'
Learn More

Shermond™ Polythene Gloves

Product Code. 12990975
Encompass_Preferred
Click to view available options
Size:
Small
Medium
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Lightweight disposable polythene gloves offering basic protection

  • Inexpensive disposable gloves provide adequate covering for light work
  • Ambidextrous
  • Unpowdered
  • Clear polythene
  • Not sterile
  • Not suitable for use with chemicals

Specifications

Certifications/Compliance None
Color Clear
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Polyethylene
Powdered Powder Free
Product Type Gloves
Size Small
Sterility Non-sterile
For Use With (Application) General Purpose use Including Food Hygiene
Glove Cuff Style Plain
Quantity 100 Pk.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'