missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Polytril™ Top Gloves
Click to view available options
Size: 9
Description
- Knitted polyamide glove conforms perfectly to the natural shape of the hand to provide excellent dexterity.
- Polyamide provides good resistance to abrasion, humidity, tearing and shearing.
- Black oil proof nitrile coating on palm, back and fingertips provides excellent abrasion resistance and oil proof properties.
- Suitable for use in the following:
- Assembly work in oily or greasy conditions.
- Outdoor handling in wet environments.
- Construction, automotive, metal industry.
- Elastic wrist.
- Black color dedicated to dirty applications and environments
Specifications
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 Basic standards, EN 388-2003 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/05/07/0190 |
Classification | Dexterity: 5 |
Coated Area | Palm, Fingers, Back |
Cuff Material | Polyamide |
Description | Gauge 13 |
Coating Material | Nitrile |
Color | Black |
Cut Resistance Rating | 1 |
Material | Polyamide |
Product Type | Gloves |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'