Learn More
Kinematica™ Polytron™ Homogenizer Stands: Safety Rings
Prevents homogenizer from moving out of position.
Brand: Kinematica™ 11045035
This item is not returnable.
View return policy
Description
The safety ring prevents uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.Specifications
Safety Positioning Ring | |
Stainless Steel | |
ST-P10/600 stand |