missing translation for 'onlineSavingsMsg'
Learn More

Navailles Polyurethane Adjustable Chair

This Polyurethane Adjustable Chair is easily adjustable and features a comfortable and airy surface. Abrasion-resistant design without staples nor seams.

Brand:  Navailles 7NG 35NHPR 02 905

317.21 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 17406182

  • £358.00 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 5-branch, reinforced polyamide base with heavy-duty nylon casters
  • Gas-lift column with height adjustment and shock absorber
  • Ergonomic seat in black KANGO™ soft polyurethane
  • Articulated backrest in black soft polyurethane, adjustable in depth and height
Specifications

Specifications

Black
No Armrests
Laboratories, Chemical Industry
265 lb.
620 to 860 mm
24.4 lb.
Heavy-duty nylon wheels
60 cm
Yes
EN 1021-1, EN 1021-2
Gas-lift column
120 kg
8 Years
11.1 kg
23.6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers