missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ Classic™ Refill Bottle

Product Code. 12920365
Click to view available options
Color:
Neon yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides excellent hearing protection and all-day comfort. 3M™ E-A-R™ Classic™ Earplugs Refill Bottle gives wearers easy access in work and leisure activities.

  • Soft, energy-absorbing foam
  • Textured surface helps keep plugs in place
  • Moisture-resistant - helps maintain consistent expansion time even in humid environments
  • Dispenser serves as a reminder to wear hearing protection and make earplugs readily available
TRUSTED_SUSTAINABILITY

Specifications

Color Neon yellow
Certifications/Compliance CE Approved
For Use With (Application) For one-touch dispenser
Pairs per Package 500 Pairs
Product Type Ear Plug Refill Bottle
Disposable Single-use
Material Polyurethane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'