missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ Polyurethane Earplug

Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.

Brand:  3M™ 7000103733

53.42 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12394749

  • £90.90 / 100 pairs
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Reduces noise levels by 28 dB CE certified EN 354
  • Easy to slide gently into your ears with slow recovery patented foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
  • 100 pairs per box
  • Available with probed feature for use with 3M™ E-A-RFit™ Dual-Ear Validation System

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yellow, Blue
28 dB
Ear Plugs
CE Approved
Uncorded
Polyurethane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ E-A-R™ Polyurethane Earplug > Color: Yellow/Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.