missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ Polyurethane Earplug

Product Code. p-8006610
Click to view available options
Color:
Orange
Yellow, Blue
Corded:
Corded
Uncorded
Noise Reduction Rating (NRR):
25 dB
28 dB
34 dB
Unit Size:
100 pairs
50 pairs
Pack of 250
Pack of 500
This item is not returnable. View return policy

Product Code. 12394749

Brand: 3M™ 7000103733

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.

  • Reduces noise levels by 28 dB CE certified EN 354
  • Easy to slide gently into your ears with slow recovery patented foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
  • 100 pairs per box
  • Available with probed feature for use with 3M™ E-A-RFit™ Dual-Ear Validation System

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow, Blue
Certifications/Compliance CE Approved
Noise Reduction Rating (NRR) 28 dB
Corded Uncorded
Product Type Ear Plugs
Material Polyurethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.