missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polyurethane Foam Stoppers
Formed by polymerization in the presence of a foaming agent.
Brand: Fisherbrand™
Code : 02
Additional Details : Weight : 0.06300kg
Description
- Made up of polyurethane
- Rigid and flexible
Spécification
Plug Stopper | |
30 mm | |
Polyurethane |
40 mm | |
100/Pk. |