missing translation for 'onlineSavingsMsg'
Learn More
Polyvinylpyrrolidone, average M.W. 50.000, K30, Thermo Scientific Chemicals Product Code.: 10721701

Polyvinylpyrrolidone, average M.W. 50.000, K30, Thermo Scientific Chemicals

Product Code. 10721701
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Pail
Plastic bottle
This item is not returnable. View return policy

Product Code. 10721701

Brand: Thermo Scientific Acros 227541000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidoneShow MoreShow Less
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
pH 3.0 to 5.0 (5% aq. soln.)
Physical Form Crystalline Powder or Crystals
Melting Point 225.0°C
Avg. Mol. Wt. or Mol. Wt. Range 50.000
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 100 g
Percent Purity 11.5 to 12.8% (Nitrogen, on anh. subst.) 10ppm 1-vinylpyrrolidin-2-one (HPLC) max. 3% max. 2-pyrrolidone (HPLC)
Identification B,C,D,E comply
Infrared Spectrum Authentic
Packaging Plastic bottle
Viscosity 29 to 32 K−value (1% solid w/v aq. soln.)
Water 5% Max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, K29-32
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466382 Certificate TypeCertificate of Analysis Date04/10/2024 Catalog Number227545000,227541000,227540025
Lot NumberA0463979 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number227545000,227541000,227540025
Lot NumberA0457919 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number227545000,227541000,227540025
Lot NumberA0454256 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number227545000,227541000,227540025
Lot NumberA0393790 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number227545000,227541000,227540025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyvinylpyrrolidone, average M.W. 50.000, K30, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.