Learn More
Polyvinylpyrrolidone, average M.W. 8.000, K16-18, Thermo Scientific Chemicals
£60.00 - £204.02
Chemical Identifiers
CAS | 9003-39-8 |
---|---|
Molecular Formula | (C6H9NO)n |
Molecular Weight (g/mol) | 111.14 |
MDL Number | MFCD01076626 |
InChI Key | WHNWPMSKXPGLAX-UHFFFAOYSA-N |
Synonym | n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone |
PubChem CID | 6917 |
ChEBI | CHEBI:82551 |
IUPAC Name | 1-ethenylpyrrolidin-2-one |
SMILES | *-CC(-*)N1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10134760
|
Thermo Scientific Acros
227465000 |
500 g | Plastic bottle |
£60.00
500g |
In Stock
Log in to see stock. |
||||
10114333
|
Thermo Scientific Acros
227460025 |
2.5 kg | Plastic bottle |
£204.02
2.50kg |
Estimated Shipment: 03-01-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9003-39-8 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
(C6H9NO)n | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
*-CC(-*)N1CCCC1=O |
Specifications
White or White-Yellow | |
9003-39-8 | |
130.0°C | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
6917 | |
>215°C | |
Positive | |
5% Max. (K.F.) |
3.0 to 5.0 (5% aq. soln.) | |
Powder or Flakes | |
(C6H9NO)n | |
15, 7814 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether | |
*-CC(-*)N1CCCC1=O | |
111.14 | |
CHEBI:82551 | |
11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC) | |
Authentic | |
Polyvinylpyrrolidone, K16-18 |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
missing translation for 'rtecsNumber' : TR8250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.