missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polywipe-C Wipes
GREENER_CHOICE

Reduce solvent usage and maximize process control and consistency in general purpose and critical cleaning applications. Contec™ Polywipe-C Wipes are saturated with 100% IPA or a 70% isopropanol/30% deionized water solution.

Brand:  Contec™ PSCS0012

 View more versions of this product

Product Code. 16655534

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lightweight wipes have laser sealed edges and are laundered and saturated in Contec's ISO Class 4 environment for the lowest levels of particles, fibers and extractable residue
  • Textured wipe is effective, yet cleans surfaces without scratching
  • Made from 100% textured knitted polyester
  • Saturated with 100% IPA or a 70% isopropanol/30% deionized water solution
  • Reduce VOCs associated with IPA
  • Resealable pouch preserves cleanliness and solvent saturation levels
Specifications

Specifications

Wipe
30 cm
30 cm
Single-use
Ideal for cleaning tools, instruments and other equipment
70% IPA/30% DI water
12 in.
12 in.
Not Autoclavable
30 x 30 cm (12 x 12 in.)
1-ply
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers