missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polywipe-C Wipes
BIOPHARMA

Reduce solvent usage and maximize process control and consistency in general purpose and critical cleaning applications. Contec™ Polywipe-C Wipes are saturated with 100% IPA or a 70% isopropanol/30% deionized water solution.

Brand:  Contec™ PSCS0012-BPR

699.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 17284523

  • £902.00 / Pack of 480

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lightweight wipes have laser sealed edges and are laundered and saturated in Contec's ISO Class 4 environment for the lowest levels of particles, fibers and extractable residue
  • Textured wipe is effective, yet cleans surfaces without scratching
  • Made from 100% textured knitted polyester
  • Saturated with 100% IPA or a 70% isopropanol/30% deionized water solution
  • Reduce VOCs associated with IPA
  • Resealable pouch preserves cleanliness and solvent saturation levels
Specifications

Specifications

Wipe
30 cm
30 cm
Not Autoclavable
30 x 30 cm (12 x 12 in.)
1-ply
Non-sterile
12 in.
12 in.
Knitted Polyester
Single-use
Ideal for Cleaning Tools, Instruments and Other Equipment
70% IPA/30% DI Water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers