missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POM 1, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Inhibitor of Ecto-NTPDases that displays minor selectivity for NTPDases 1 and 2 over NTPDase 3 and P2Y12 (Ki values are 2.58, 3.26, > 10 and 28.8 μM for NTPDase 1, NTPDase 3, P2Y12 and NTPDase 2 respectively). Abolishes renal protection induced by ischemic preconditioning. Inhibits synaptic transmission at hippocampal CA1 pyramidal synapses and at the cerebellar parallel fiber-Purkinje cell (PF) synapse.
Specifications
Specifications
CAS | 12141-67-2 |
Quantity | 50 mg |
Molecular Formula | H2O40Na6W12 |
Synonym | Sodium metatungstate |
Formula Weight | 2986.01 |
Percent Purity | >98% |
Chemical Name or Material | POM 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction