Learn More
Azlon™ POM Autoclavable Tweezer
Polyoxymethylene (POM) construction. Azlon™ POM Autoclavable Tweezers are suitable for multiple applications.
Brand:  Azlon TWY264
9.96 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12033984
  • £16.30 / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polyoxymethylene (POM) construction. Azlon™ POM Autoclavable Tweezers are suitable for multiple applications.
  • High quality, autoclavable tweezers
  • Rounded ends
  • Fine tooth grips for secure handling of samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round
POM
Tweezers
180 L mm
7.09 in.
18.0 cm
For secure handling of samples
Ea.