Learn More
Kartell™ POM Slide Staining Trough
Stain up to 25 slides with a versatile trough that can also be used as a mailing box. Kartell™ POM Slide Staining Trough is constructed of POM and designed to hold up to 25 slides.
Brand:  Kartell™ 0110100
Product Code. 10395961
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Stain up to 25 slides with a versatile trough that can also be used as a mailing box. Kartell™ POM Slide Staining Trough is constructed of POM and designed to hold up to 25 slides.
  • Holds up to 25 slides
  • Accommodates one Kartell™ 1100 rack
  • Can also be used as a mailing box
Specifications
Specifications
Staining Trough
Slides
52.4 mm
25
88 x 100.8 x 52.4 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Kartell™ POM Slide Staining Trough > Height: 52.4mm