missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pomalidomide

Product Code. 16524504 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16524504

Brand: Thermo Scientific Chemicals 465700010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19171-19-8 | C13H11N3O4 | 273.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD12756407
InChI Key UVSMNLNDYGZFPF-UHFFFAOYNA-N
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O

Specifications

Melting Point 315.0°C to 320.0°C
Quantity 1 g
Formula Weight 273.24
Chemical Name or Material Pomalidomide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.