Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG1-azide
Cereblon ligand with PEG linker and terminal azide for onward chemistry
Brand: Tocris Bioscience™ 7434/25
Additional Details : Weight : 0.00970kg
Description
Functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
4-[[2-(2-Azidoethoxy)ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione | |
25 mg | |
C17H18N6O5 |
2271036-44-1 | |
Degrader Building Blocks | |
0.95 |