missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ponceau S Staining Solution

Product Code. 17641536 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
1000 mL
3 x 1000 mL
Unit Size:
1000mL
500mL
Pack of 3
This item is not returnable. View return policy

Product Code. 17641536

Brand: Thermo Scientific™ A40000278

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Product Type Ponceau S Staining Solution
Form Liquid
Formulation 0.1% (w/v) in 5% acetic acid
For Use With (Application) Western Blotting
Quantity 1000 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.